Learn More
Meso Scale Discovery VPLEX Cytokine PNL 1 human Kit
SDP
Catalog No. 5000078224
Encompass
The Meso Scale Discovery V-PLEX Human Cytokine Panel 1 Kit is a multiplex immunoassay that simultaneously measures 10 human cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from cell culture supernatants serum plasma and other biological fluids The assay utilizes MSD s electrochemiluminescent detection technology and is run on MSD s QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and diluents valid for 50 tests The assay has high sensitivity broad dynamic range and requires low sample volumes
Catalog No. 50-000-78224 Supplier Meso Scale Discovery Supplier No. K15050D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human Cytokine Panel 1 Kit is a multiplex immunoassay that simultaneously measures 10 human cytokines and chemokines (IFN- IL-10 IL-12p70 IL-13 IL-1 IL-2 IL-4 IL-6 IL-8 TNF- ) from cell culture supernatants serum plasma and other biological fluids The assay utilizes MSD s electrochemiluminescent detection technology and is run on MSD s QuickPlex SQ 120 instrument The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and diluents valid for 50 tests The assay has high sensitivity broad dynamic range and requires low sample volumes