Learn More
Meso Scale Discovery VPLEX Cytokine PNL 1 mouse
SDP
Catalog No. 5000081326
Encompass
The Meso Scale Discovery V-PLEX Cytokine Panel 1 (Mouse) Kit is a multiplex immunoassay that simultaneously measures mouse IFN- IL-10 IL-12p70 IL-1 IL-2 IL-4 IL-5 IL-6 KC/GRO and TNF- in serum plasma and culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals sufficient for one 96-well plate This assay allows for the accurate quantitation of multiple cytokines from a small sample volume
Catalog No. 50-000-81326 Supplier Meso Scale Discovery Supplier No. K15245D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Cytokine Panel 1 (Mouse) Kit is a multiplex immunoassay that simultaneously measures mouse IFN- IL-10 IL-12p70 IL-1 IL-2 IL-4 IL-5 IL-6 KC/GRO and TNF- in serum plasma and culture supernatants It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals sufficient for one 96-well plate This assay allows for the accurate quantitation of multiple cytokines from a small sample volume