Learn More
Meso Scale Discovery VPLEX Cytokine PNL 1 NHP Kit
SDP
Catalog No. 5000081758
Encompass
The V-PLEX NHP Cytokine Panel 1 Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring 10 non-human primate cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IFN- TNF- ) in serum plasma and culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies assay diluent wash buffer read buffer and calibrator enabling efficient and accurate NHP cytokine profiling
Catalog No. 50-000-81758 Supplier Meso Scale Discovery Supplier No. K15057D2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX NHP Cytokine Panel 1 Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring 10 non-human primate cytokines and chemokines (IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IFN- TNF- ) in serum plasma and culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies assay diluent wash buffer read buffer and calibrator enabling efficient and accurate NHP cytokine profiling