Learn More
Meso Scale Discovery VPLEX Cytokine PNL 1 NHP Kit
SDP
Catalog No. 5000078253
Encompass
The Meso Scale Discovery V-PLEX Cytokine Panel 1 (NHP) Kit is designed to simultaneously measure multiple monkey cytokines in serum plasma and culture supernatants This kit can quantitatively measure IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A IFN- and TNF- The electrochemiluminescent detection method provides high sensitivity up to 5-logs of dynamic range and low sample consumption This kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer calibrator blend serum and plasma diluent wash buffer and adhesive plate seals sufficient for one 96-well plate
Catalog No. 50-000-78253 Supplier Meso Scale Discovery Supplier No. K15057D1
The Meso Scale Discovery V-PLEX Cytokine Panel 1 (NHP) Kit is designed to simultaneously measure multiple monkey cytokines in serum plasma and culture supernatants This kit can quantitatively measure IL-1 IL-2 IL-4 IL-5 IL-6 IL-10 IL-12p70 IL-13 IL-17A IFN- and TNF- The electrochemiluminescent detection method provides high sensitivity up to 5-logs of dynamic range and low sample consumption This kit includes a 96-well plate coated with capture antibodies detection antibodies read buffer calibrator blend serum and plasma diluent wash buffer and adhesive plate seals sufficient for one 96-well plate