Learn More
Meso Scale Discovery VPLEX GIP active Kit 25 PL
SDP
Catalog No. 5000078718
Encompass
The Meso Scale Discovery V-PLEX GIP Active Kit is a 96-well plate assay that measures active glucagon-like peptide-1 (GLP-1) in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 50 L The V-PLEX GIP Active Kit allows for the quantitative determination of active GIP levels and is designed for research use only not for use in diagnostic procedures The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and positive control
Catalog No. 50-000-78718 Supplier Meso Scale Discovery Supplier No. K150T5D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX GIP Active Kit is a 96-well plate assay that measures active glucagon-like peptide-1 (GLP-1) in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume of 50 L The V-PLEX GIP Active Kit allows for the quantitative determination of active GIP levels and is designed for research use only not for use in diagnostic procedures The kit includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer blocker reagents calibrator and positive control