Learn More
Meso Scale Discovery VPLEX GLP1 Active Kit 25 PL
SDP
Catalog No. 5000081568
Encompass
The Meso Scale Discovery V-PLEX GLP-1 Active Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a GLP-1 specific capture antibody a detection antibody conjugated to an electrochemiluminescent label calibrators controls assay diluent read buffer wash buffer and plate seals The assay has a range of 1 56 - 400 pg/mL and takes approximately 3 5 hours to complete This highly sensitive kit allows for the direct quantitation of active GLP-1 without sample pretreatment
Catalog No. 50-000-81568 Supplier Meso Scale Discovery Supplier No. K1503OD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX GLP-1 Active Kit is an electrochemiluminescent immunoassay for the quantitative measurement of active GLP-1 in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with a GLP-1 specific capture antibody a detection antibody conjugated to an electrochemiluminescent label calibrators controls assay diluent read buffer wash buffer and plate seals The assay has a range of 1 56 - 400 pg/mL and takes approximately 3 5 hours to complete This highly sensitive kit allows for the direct quantitation of active GLP-1 without sample pretreatment