Learn More
Meso Scale Discovery VPLEX GLP1 Total Kit 25 PL
SDP
Catalog No. 5000081820
Encompass
The Meso Scale Discovery V-PLEX GLP-1 Total Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range and rapid protocol This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The lower limit of detection is 1 5 pg/mL with an upper limit of 10 000 pg/mL
Catalog No. 50-000-81820 Supplier Meso Scale Discovery Supplier No. K1503PD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX GLP-1 Total Kit is an electrochemiluminescent immunoassay for the quantitative measurement of total glucagon-like peptide-1 (GLP-1) in human serum plasma and cell culture supernatants The assay utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with a broad dynamic range and rapid protocol This kit includes a 96-well plate coated with capture antibody detection antibody conjugated to ruthenium label calibrators and reagents required to perform the assay The lower limit of detection is 1 5 pg/mL with an upper limit of 10 000 pg/mL