Learn More
Meso Scale Discovery VPLEX HUM Biomarker 39Plex Kit
SDP
Catalog No. 5000080489
Encompass
The V-PLEX Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows simultaneous quantification of 40 biomarkers in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies for the 40 analytes detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The assay uses electrochemiluminescence for high sensitivity and broad dynamic range This kit allows efficient analysis of biomarkers related to inflammation immune response angiogenesis tissue remodeling and more in one experiment from a small sample volume
Catalog No. 50-000-80489 Supplier Meso Scale Discovery Supplier No. K15209D1
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay that allows simultaneous quantification of 40 biomarkers in human serum plasma and cell culture supernatants The kit contains a 96-well plate coated with capture antibodies for the 40 analytes detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The assay uses electrochemiluminescence for high sensitivity and broad dynamic range This kit allows efficient analysis of biomarkers related to inflammation immune response angiogenesis tissue remodeling and more in one experiment from a small sample volume