Learn More
Meso Scale Discovery VPLEX HUM Biomarker 39Plex
SDP
Catalog No. 5000080113
Encompass
The V-PLEX Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay for quantifying 40 biomarkers in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and low sample volume The panel includes cytokines chemokines growth factors soluble receptors and cardiovascular markers Suitable for biomarker discovery translational research clinical research and diagnostics the kit includes a 96-well plate detection antibodies calibrators diluents wash buffers readout buffer and plate seals
Catalog No. 50-000-80113 Supplier Meso Scale Discovery Supplier No. K15209D4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human Biomarker 40-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay for quantifying 40 biomarkers in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity broad dynamic range and low sample volume The panel includes cytokines chemokines growth factors soluble receptors and cardiovascular markers Suitable for biomarker discovery translational research clinical research and diagnostics the kit includes a 96-well plate detection antibodies calibrators diluents wash buffers readout buffer and plate seals