Learn More
Meso Scale Discovery VPLEX HUM CPeptide Kit 25 PL
SDP
Catalog No. 5000081486
Encompass
The Meso Scale Discovery V-PLEX Human C-Peptide Kit is a multiplex immunoassay that quantitatively measures C-peptide levels in human serum plasma and culture supernatant samples The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer and wash buffer The electrochemiluminescent assay has a detection range of 50-10 000 pg/mL and requires 25 L of sample The total assay time is approximately 3 5 hours This highly sensitive and reproducible kit can monitor C-peptide levels and -cell function
Catalog No. 50-000-81486 Supplier Meso Scale Discovery Supplier No. K151X5D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human C-Peptide Kit is a multiplex immunoassay that quantitatively measures C-peptide levels in human serum plasma and culture supernatant samples The kit contains a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer and wash buffer The electrochemiluminescent assay has a detection range of 50-10 000 pg/mL and requires 25 L of sample The total assay time is approximately 3 5 hours This highly sensitive and reproducible kit can monitor C-peptide levels and -cell function