Learn More
Meso Scale Discovery VPLEX HUM Cytokine 29Plex
SDP
Catalog No. 5000081823
Encompass
The V-PLEX Human Cytokine 30-plex Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures 30 human cytokines and chemokines in a single small sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls buffers and reagents for quantitative analysis of cytokines including IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IL-10 IL-12p70 IL-13 IL-15 IL-16 IL-17A TNF- IFN- GM-CSF VEGF Eotaxin IP-10 MCP-1 MIP-1 MIP-1 RANTES EGF FGF-2 G-CSF HGF and PDGF-BB The kit utilizes MSD s electrochemiluminescent detection technology for high sensitivity and broad dynamic range
Catalog No. 50-000-81823 Supplier Meso Scale Discovery Supplier No. K15054D4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human Cytokine 30-plex Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures 30 human cytokines and chemokines in a single small sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls buffers and reagents for quantitative analysis of cytokines including IL-1 IL-2 IL-4 IL-5 IL-6 IL-7 IL-8 IL-10 IL-12p70 IL-13 IL-15 IL-16 IL-17A TNF- IFN- GM-CSF VEGF Eotaxin IP-10 MCP-1 MIP-1 MIP-1 RANTES EGF FGF-2 G-CSF HGF and PDGF-BB The kit utilizes MSD s electrochemiluminescent detection technology for high sensitivity and broad dynamic range