Learn More
Meso Scale Discovery VPLEX HUM GMCSF Kit 25Plate
SDP
Catalog No. 5000079177
Encompass
The V-PLEX Human GM-CSF Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring GM-CSF in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range requiring low sample volume The assay quantifies natural and recombinant GM-CSF with a detection range of 0 10 to 10 000 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrator diluents for a standard curve
Catalog No. 50-000-79177 Supplier Meso Scale Discovery Supplier No. K151RID4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human GM-CSF Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring GM-CSF in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection for high sensitivity and broad dynamic range requiring low sample volume The assay quantifies natural and recombinant GM-CSF with a detection range of 0 10 to 10 000 pg/mL The kit includes a 96-well plate with capture antibodies detection antibodies with electrochemiluminescent labels and calibrator diluents for a standard curve