Learn More
Meso Scale Discovery VPLEX HUM IL12/IL23p40 Kit
SDP
Catalog No. 5000078763
Encompass
The Meso Scale Discovery V-PLEX Human IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for quantifying human IL-12 and IL-23 in serum plasma and culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager offering high sensitivity and a wide dynamic range for simple rapid cytokine measurement
Catalog No. 50-000-78763 Supplier Meso Scale Discovery Supplier No. K151RJD1
The Meso Scale Discovery V-PLEX Human IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for quantifying human IL-12 and IL-23 in serum plasma and culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents Results are analyzed on MSD s SECTOR Imager offering high sensitivity and a wide dynamic range for simple rapid cytokine measurement