Learn More
Meso Scale Discovery VPLEX HUM IL12/IL23p4025Plate
SDP
Catalog No. 5000081682
Encompass
The Meso Scale Discovery V-PLEX Human IL-12/IL-23p40 Kit is a multiplex immunoassay for measuring IL-12p40 and IL-23 in human serum plasma and culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for simultaneous detection of two analytes from one sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents It has a detection range of 0 05-1000 pg/mL and minimal cross-reactivity enabling sensitive and accurate quantification of IL-12 and IL-23 cytokines
Catalog No. 50-000-81682 Supplier Meso Scale Discovery Supplier No. K151RJD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human IL-12/IL-23p40 Kit is a multiplex immunoassay for measuring IL-12p40 and IL-23 in human serum plasma and culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for simultaneous detection of two analytes from one sample The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators and necessary reagents It has a detection range of 0 05-1000 pg/mL and minimal cross-reactivity enabling sensitive and accurate quantification of IL-12 and IL-23 cytokines