Learn More
Meso Scale Discovery VPLEX HUM IL16 Kit 25Plate
SDP
Catalog No. 5000078565
Encompass
The V-PLEX Human IL-16 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 16 (IL-16) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody to human IL-16 a detection antibody conjugated to an ECL label calibrators and reagents required for the assay The kit has a detection range of 3 2-10 000 pg/mL and exhibits no cross-reactivity with other cytokines This assay allows sensitive accurate and rapid quantification of IL-16 levels with as little as 25 L of sample
Catalog No. 50-000-78565 Supplier Meso Scale Discovery Supplier No. K151RED4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human IL-16 Kit from Meso Scale Discovery is an immunoassay that uses electrochemiluminescence (ECL) for the quantitative measurement of human interleukin 16 (IL-16) in cell culture supernatants serum plasma and other biological fluids The kit contains a 96-well plate coated with a monoclonal antibody to human IL-16 a detection antibody conjugated to an ECL label calibrators and reagents required for the assay The kit has a detection range of 3 2-10 000 pg/mL and exhibits no cross-reactivity with other cytokines This assay allows sensitive accurate and rapid quantification of IL-16 levels with as little as 25 L of sample