Learn More
Meso Scale Discovery VPLEX HUM IL1a Kit 25Plate
SDP
Catalog No. 5000078839
Encompass
The V-PLEX Human IL-1 Kit from Meso Scale Discovery is a sensitive immunoassay for measuring human interleukin 1 alpha (IL-1 ) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for accurate reproducible results with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s platforms and analysis software aids data reduction ensuring precise IL-1 quantification
Catalog No. 50-000-78839 Supplier Meso Scale Discovery Supplier No. K151RBD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human IL-1 Kit from Meso Scale Discovery is a sensitive immunoassay for measuring human interleukin 1 alpha (IL-1 ) in various biological fluids It uses MSD s SULFO-TAG labels and multi-array technology for accurate reproducible results with high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are read on MSD s platforms and analysis software aids data reduction ensuring precise IL-1 quantification