Learn More
Meso Scale Discovery VPLEX HUM IL1RA Kit 25 PL
SDP
Catalog No. 5000081799
Encompass
The V-PLEX Human IL-1ra Kit from Meso Scale Discovery is an immunoassay for measuring human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s systems providing a quantitative read-out in pg/mL offering a sensitive method for IL-1ra measurement in research
Catalog No. 50-000-81799 Supplier Meso Scale Discovery Supplier No. K151WTD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human IL-1ra Kit from Meso Scale Discovery is an immunoassay for measuring human interleukin-1 receptor antagonist (IL-1ra) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for high sensitivity and low sample volume The kit includes a 96-well plate with capture antibody detection antibody with ruthenium label calibrators and necessary reagents Results are analyzed on MSD s systems providing a quantitative read-out in pg/mL offering a sensitive method for IL-1ra measurement in research