Learn More
Meso Scale Discovery VPLEX HUM Insulin Kit 25 PL
SDP
Catalog No. 5000081297
Encompass
The V-PLEX Human Insulin Kit from Meso Scale Discovery is a multiplex immunoassay for measuring insulin in human serum plasma saliva and cell culture samples It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators and necessary reagents The detection range is 50-25 000 pg/mL with minimal proinsulin cross-reactivity The assay takes about 3 5 hours and requires an MSD instrument like QuickPlex SECTOR Imager or SQ 120 It offers a sensitive specific and high-throughput method for insulin measurement
Catalog No. 50-000-81297 Supplier Meso Scale Discovery Supplier No. K151S5D4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human Insulin Kit from Meso Scale Discovery is a multiplex immunoassay for measuring insulin in human serum plasma saliva and cell culture samples It includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators and necessary reagents The detection range is 50-25 000 pg/mL with minimal proinsulin cross-reactivity The assay takes about 3 5 hours and requires an MSD instrument like QuickPlex SECTOR Imager or SQ 120 It offers a sensitive specific and high-throughput method for insulin measurement