Learn More
Meso Scale Discovery VPLEX HUM Leptin Kit 25 PL
SDP
Catalog No. 5000078527
Encompass
The Meso Scale Discovery V-PLEX Human Leptin Kit is a sensitive immunoassay for detecting human leptin in serum plasma and cell culture supernatants It uses electrochemiluminescent technology delivering results in 3 5 hours The kit includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals It has a sensitivity of 19 pg/mL and a range of 19-10 000 pg/mL allowing precise leptin measurement The kit tests 39 samples in duplicate with calibrators and controls
Catalog No. 50-000-78527 Supplier Meso Scale Discovery Supplier No. K151V5D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Human Leptin Kit is a sensitive immunoassay for detecting human leptin in serum plasma and cell culture supernatants It uses electrochemiluminescent technology delivering results in 3 5 hours The kit includes a 96-well plate with capture antibody detection antibody with an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals It has a sensitivity of 19 pg/mL and a range of 19-10 000 pg/mL allowing precise leptin measurement The kit tests 39 samples in duplicate with calibrators and controls