Learn More
Meso Scale Discovery VPLEX HUM Prinflm PNL 4PlxPlt
SDP
Catalog No. 5000078275
Encompass
The Meso Scale Discovery V-PLEX Plus Human ProInflam Panel I 4-plex is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 4 human proinflammatory biomarkers (IFN- IL-1 IL-6 and TNF- ) from cell culture supernatants serum plasma and other biological samples It utilizes MSD s electrochemiluminescent detection technology for high sensitivity wide dynamic range and minimal matrix interference The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and other reagents required to perform the assay It provides a simple rapid and robust tool for inflammation-related research
Catalog No. 50-000-78275 Supplier Meso Scale Discovery Supplier No. K15052G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Human ProInflam Panel I 4-plex is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 4 human proinflammatory biomarkers (IFN- IL-1 IL-6 and TNF- ) from cell culture supernatants serum plasma and other biological samples It utilizes MSD s electrochemiluminescent detection technology for high sensitivity wide dynamic range and minimal matrix interference The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and other reagents required to perform the assay It provides a simple rapid and robust tool for inflammation-related research