Learn More
Meso Scale Discovery VPLEX HUM Prinflm PNL 4PlxPlt
SDP
Catalog No. 5000081570
Encompass
The V-PLEX Plus Human ProInflam Panel I 4-plex Kit from Meso Scale Discovery is a multiplex immunoassay measuring human IFN- IL-1 IL-6 and TNF- in serum plasma and culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results from small samples The 4-plex panel efficiently quantifies proinflammatory cytokines aiding immunology inflammation and drug discovery research The kit includes a 96-well plate detection antibodies read buffer calibrators and protocols for assay optimization and data analysis
Catalog No. 50-000-81570 Supplier Meso Scale Discovery Supplier No. K15052G1
The V-PLEX Plus Human ProInflam Panel I 4-plex Kit from Meso Scale Discovery is a multiplex immunoassay measuring human IFN- IL-1 IL-6 and TNF- in serum plasma and culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive reproducible results from small samples The 4-plex panel efficiently quantifies proinflammatory cytokines aiding immunology inflammation and drug discovery research The kit includes a 96-well plate detection antibodies read buffer calibrators and protocols for assay optimization and data analysis