Learn More
Meso Scale Discovery VPLEX HUM Prnflm PNL II 4PlexP
SDP
Catalog No. 5000079192
Encompass
The Meso Scale Discovery V-PLEX Plus Human ProInflam Panel II 4-plex is a kit for measuring 4 human proinflammatory biomarkers (IFN- IL-1 IL-6 and TNF- ) in serum plasma and cell culture supernatants using electrochemiluminescence detection technology The kit includes a 4-spot MULTI-ARRAY 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3-0 8 pg/mL and a 3 5 hour total assay time This multiplex immunoassay allows simultaneous quantification of multiple biomarkers from a single small sample volume
Catalog No. 50-000-79192 Supplier Meso Scale Discovery Supplier No. K15053G1
The Meso Scale Discovery V-PLEX Plus Human ProInflam Panel II 4-plex is a kit for measuring 4 human proinflammatory biomarkers (IFN- IL-1 IL-6 and TNF- ) in serum plasma and cell culture supernatants using electrochemiluminescence detection technology The kit includes a 4-spot MULTI-ARRAY 96-well plate coated with capture antibodies detection antibodies calibrators diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3-0 8 pg/mL and a 3 5 hour total assay time This multiplex immunoassay allows simultaneous quantification of multiple biomarkers from a single small sample volume