Learn More
Meso Scale Discovery VPLEX HUM TNFSY Kit 25Plate
SDP
Catalog No. 5000082219
Encompass
The V-PLEX Human TNF-beta Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human TNF-beta in various biological fluids It uses electrochemiluminescence detection with high sensitivity up to 5 pg/mL in an antibody sandwich format The kit includes a 96-well plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions The assay takes about 3 hours and requires a Meso Scale Discovery instrument allowing rapid accurate quantification with minimal sample volume and hands-on time
Catalog No. 50-000-82219 Supplier Meso Scale Discovery Supplier No. K151RGD4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Human TNF-beta Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human TNF-beta in various biological fluids It uses electrochemiluminescence detection with high sensitivity up to 5 pg/mL in an antibody sandwich format The kit includes a 96-well plate with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions The assay takes about 3 hours and requires a Meso Scale Discovery instrument allowing rapid accurate quantification with minimal sample volume and hands-on time