Learn More
Meso Scale Discovery VPLEX HUM VEGF Kit 25Plate
SDP
Supplier:  Meso Scale Discovery K151RHD4
Encompass
The Meso Scale Discovery V-PLEX Human VEGF Kit is a multiplex immunoassay that measures vascular endothelial growth factor (VEGF) in human serum plasma and cell culture supernatants The kit uses electrochemiluminescence (ECL) detection technology and provides high sensitivity a broad dynamic range and requires low sample volumes The V-PLEX VEGF Kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with ECL tags calibrators diluent wash buffer read buffer and plate seals The assay has a typical sensitivity of 1 3 pg/mL and an upper limit of quantification of 10 000 pg/mL for VEGF
Catalog No. 50-000-79795
Add to Cart
Request a Quote