Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1 canine
SDP
Catalog No. 5000078316
Encompass
The V-PLEX Metabolic Panel 1 Canine Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring albumin alkaline phosphatase alanine aminotransferase amylase creatinine glucose total calcium phosphorus total protein blood urea nitrogen sodium potassium and chloride in canine serum plasma and urine It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The electrochemiluminescent immunoassay offers high sensitivity and a broad dynamic range enabling efficient assessment of canine metabolism and organ function from a small sample
Catalog No. 50-000-78316 Supplier Meso Scale Discovery Supplier No. K15335D4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Metabolic Panel 1 Canine Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring albumin alkaline phosphatase alanine aminotransferase amylase creatinine glucose total calcium phosphorus total protein blood urea nitrogen sodium potassium and chloride in canine serum plasma and urine It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The electrochemiluminescent immunoassay offers high sensitivity and a broad dynamic range enabling efficient assessment of canine metabolism and organ function from a small sample