Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1 human
SDP
Catalog No. 5000079846
Encompass
The V-PLEX Metabolic Panel 1 Human Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 10 metabolic biomarkers from a single sample The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The 10 biomarkers measured are albumin total protein creatinine glucose AST ALT ALP total bilirubin BUN and CRP The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit allows efficient metabolic testing from small sample volumes
Catalog No. 50-000-79846 Supplier Meso Scale Discovery Supplier No. K15325D4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Metabolic Panel 1 Human Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 10 metabolic biomarkers from a single sample The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent wash buffer read buffer and plate seals The 10 biomarkers measured are albumin total protein creatinine glucose AST ALT ALP total bilirubin BUN and CRP The electrochemiluminescent detection method provides high sensitivity and a broad dynamic range This kit allows efficient metabolic testing from small sample volumes