Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1 mouse
SDP
Catalog No. 5000080351
Encompass
The V-PLEX Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring 10 metabolic biomarkers in mouse serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The analytes are albumin total cholesterol HDL LDL triglycerides glucose creatinine blood urea nitrogen phosphorus and calcium The electrochemiluminescent detection offers high sensitivity and a broad dynamic range enabling efficient metabolic profiling of mouse models
Catalog No. 50-000-80351 Supplier Meso Scale Discovery Supplier No. K15333D4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring 10 metabolic biomarkers in mouse serum plasma and cell culture supernatants It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals The analytes are albumin total cholesterol HDL LDL triglycerides glucose creatinine blood urea nitrogen phosphorus and calcium The electrochemiluminescent detection offers high sensitivity and a broad dynamic range enabling efficient metabolic profiling of mouse models