Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1 NHP Kit
SDP
Catalog No. 5000079849
Encompass
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Kit quantifies insulin leptin adiponectin resistin and C-peptide in non-human primate samples Using MSD s V-PLEX technology it offers high sensitivity and rapid results The kit includes a 96-well plate with capture and detection antibodies calibrators and controls for standard curves This assay efficiently analyzes metabolic biomarkers for diabetes obesity and metabolic syndrome research requiring minimal sample volume and providing a streamlined workflow for metabolic phenotyping in non-human primate studies
Catalog No. 50-000-79849 Supplier Meso Scale Discovery Supplier No. K15332D2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Kit quantifies insulin leptin adiponectin resistin and C-peptide in non-human primate samples Using MSD s V-PLEX technology it offers high sensitivity and rapid results The kit includes a 96-well plate with capture and detection antibodies calibrators and controls for standard curves This assay efficiently analyzes metabolic biomarkers for diabetes obesity and metabolic syndrome research requiring minimal sample volume and providing a streamlined workflow for metabolic phenotyping in non-human primate studies