Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1 NHP Kit
SDP
Catalog No. 5000082140
Encompass
The V-PLEX Metabolic Panel 1 (NHP) Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring insulin leptin glucagon GIP PYY amylin and C-peptide in non-human primate samples It uses electrochemiluminescence for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture and detection antibodies calibrators controls diluent read and wash buffers and plate seals It supports efficient analysis of metabolic biomarkers related to diabetes obesity and metabolic syndrome
Catalog No. 50-000-82140 Supplier Meso Scale Discovery Supplier No. K15332D1
The V-PLEX Metabolic Panel 1 (NHP) Kit from Meso Scale Diagnostics is a multiplex immunoassay for measuring insulin leptin glucagon GIP PYY amylin and C-peptide in non-human primate samples It uses electrochemiluminescence for high sensitivity and a broad dynamic range with low sample volume The kit includes a 96-well plate with capture and detection antibodies calibrators controls diluent read and wash buffers and plate seals It supports efficient analysis of metabolic biomarkers related to diabetes obesity and metabolic syndrome