Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1 NHP
SDP
Catalog No. 5000078270
Encompass
The Meso Scale Discovery V-PLEX Metabolic Panel 1 (Non-Human Primate) Kit quantifies insulin leptin adiponectin resistin and C-peptide in non-human primate serum plasma and tissue culture media This ECL multiplex assay uses MSD s V-PLEX technology offering sensitive reproducible results with just 25 L of sample The kit includes a 96-well plate with capture antibodies calibrators detection antibodies read buffer wash buffer and plate seals enabling efficient analysis of metabolic biomarkers for diabetes obesity and metabolic syndrome research
Catalog No. 50-000-78270 Supplier Meso Scale Discovery Supplier No. K15332D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Metabolic Panel 1 (Non-Human Primate) Kit quantifies insulin leptin adiponectin resistin and C-peptide in non-human primate serum plasma and tissue culture media This ECL multiplex assay uses MSD s V-PLEX technology offering sensitive reproducible results with just 25 L of sample The kit includes a 96-well plate with capture antibodies calibrators detection antibodies read buffer wash buffer and plate seals enabling efficient analysis of metabolic biomarkers for diabetes obesity and metabolic syndrome research