Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1 rat Kit
SDP
Catalog No. 5000081517
Encompass
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Rat Kit is designed to measure biomarkers related to metabolism and organ health in rat serum plasma and culture media samples This multiplex immunoassay kit includes reagents to quantitatively measure insulin leptin resistin and PAI-1 The electrochemiluminescent detection method allows sensitive accurate and rapid analysis using as little as 25 L of sample This kit provides a streamlined workflow to profile metabolic status and is intended for research use only
Catalog No. 50-000-81517 Supplier Meso Scale Discovery Supplier No. K15334D1
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Rat Kit is designed to measure biomarkers related to metabolism and organ health in rat serum plasma and culture media samples This multiplex immunoassay kit includes reagents to quantitatively measure insulin leptin resistin and PAI-1 The electrochemiluminescent detection method allows sensitive accurate and rapid analysis using as little as 25 L of sample This kit provides a streamlined workflow to profile metabolic status and is intended for research use only