Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1 rat
SDP
Catalog No. 5000079389
Encompass
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Rat Kit is designed to measure biomarkers related to metabolism and organ health in rat serum plasma and culture media samples This multiplex immunoassay kit includes reagents to quantitatively measure insulin leptin resistin and PAI-1 The electrochemiluminescent detection method allows sensitive accurate and rapid analysis using as little as 25 L of sample This kit can help researchers investigate metabolic diseases obesity diabetes and other conditions in rat models With minimal sample consumption this kit enables efficient metabolic profiling during preclinical drug development and academic research
Catalog No. 50-000-79389 Supplier Meso Scale Discovery Supplier No. K15334D4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Rat Kit is designed to measure biomarkers related to metabolism and organ health in rat serum plasma and culture media samples This multiplex immunoassay kit includes reagents to quantitatively measure insulin leptin resistin and PAI-1 The electrochemiluminescent detection method allows sensitive accurate and rapid analysis using as little as 25 L of sample This kit can help researchers investigate metabolic diseases obesity diabetes and other conditions in rat models With minimal sample consumption this kit enables efficient metabolic profiling during preclinical drug development and academic research