Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1HMNKit
SDP
Catalog No. 5000079393
Encompass
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Human Kit quantifies insulin leptin resistin PAI-1 total and MCP-1 in human serum plasma and cell culture supernatants This electrochemiluminescent multiplex assay uses MSD s V-PLEX technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for standard curves and assay validation It offers a streamlined workflow for analyzing metabolic biomarkers related to obesity diabetes and cardiovascular disease
Catalog No. 50-000-79393 Supplier Meso Scale Discovery Supplier No. K15325D1
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Human Kit quantifies insulin leptin resistin PAI-1 total and MCP-1 in human serum plasma and cell culture supernatants This electrochemiluminescent multiplex assay uses MSD s V-PLEX technology for sensitive reproducible results with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators and controls for standard curves and assay validation It offers a streamlined workflow for analyzing metabolic biomarkers related to obesity diabetes and cardiovascular disease