Learn More
Meso Scale Discovery VPLEX Metabolic PNL 1HMNKit
SDP
Catalog No. 5000082212
Encompass
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Human Kit quantifies insulin leptin resistin PAI-1 total and MCP-1 in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples It includes a 96-well plate with capture antibodies SULFO-TAG detection antibodies diluents calibrators and controls enabling accurate measurement of biomarkers related to obesity diabetes cardiovascular disease and metabolic disorders
Catalog No. 50-000-82212 Supplier Meso Scale Discovery Supplier No. K15325D2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Metabolic Panel 1 Human Kit quantifies insulin leptin resistin PAI-1 total and MCP-1 in human serum plasma and cell culture supernatants This electrochemiluminescent assay uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results from small samples It includes a 96-well plate with capture antibodies SULFO-TAG detection antibodies diluents calibrators and controls enabling accurate measurement of biomarkers related to obesity diabetes cardiovascular disease and metabolic disorders