Learn More
Meso Scale Discovery VPLEX Metabolic PNL canine
SDP
Catalog No. 5000078315
Encompass
The V-PLEX Metabolic Panel 1 Canine Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring canine metabolic biomarkers The kit contains reagents for 96 tests and can quantitatively measure albumin alkaline phosphatase alanine aminotransferase amylase creatinine glucose and total protein from a single small sample volume The electrochemiluminescent detection method allows for high sensitivity and a broad dynamic range This product would be useful for veterinary diagnostic labs running canine blood chemistry panels
Catalog No. 50-000-78315 Supplier Meso Scale Discovery Supplier No. K15335D2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Metabolic Panel 1 Canine Kit from Meso Scale Diagnostics is a multiplex immunoassay kit for measuring canine metabolic biomarkers The kit contains reagents for 96 tests and can quantitatively measure albumin alkaline phosphatase alanine aminotransferase amylase creatinine glucose and total protein from a single small sample volume The electrochemiluminescent detection method allows for high sensitivity and a broad dynamic range This product would be useful for veterinary diagnostic labs running canine blood chemistry panels