Learn More
Meso Scale Discovery VPLEX Metabolic PNL mouse
SDP
Catalog No. 5000079842
Encompass
The V-PLEX Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 10 metabolic biomarkers in mouse serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies for leptin insulin GLP-1 PAI-1 resistin MCP-1 IL-6 TNF-alpha IFN-gamma and IL-10 It utilizes electrochemiluminescence detection and provides high sensitivity broad dynamic range and requires low sample volume This kit is designed for research use only to study metabolic diseases diabetes inflammation and immunology using mouse samples
Catalog No. 50-000-79842 Supplier Meso Scale Discovery Supplier No. K15333D2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Metabolic Panel 1 Mouse Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous quantitative measurement of 10 metabolic biomarkers in mouse serum plasma and cell culture supernatants The kit includes a 96-well plate coated with capture antibodies for leptin insulin GLP-1 PAI-1 resistin MCP-1 IL-6 TNF-alpha IFN-gamma and IL-10 It utilizes electrochemiluminescence detection and provides high sensitivity broad dynamic range and requires low sample volume This kit is designed for research use only to study metabolic diseases diabetes inflammation and immunology using mouse samples