Learn More
Meso Scale Discovery VPLEX Mouse IL27p28/IL30 Kit
SDP
Catalog No. 5000078691
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-27p28/IL-30 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-27p28 and IL-30 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is sensitive reproducible and requires a low sample volume of 25 L The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating standard curves The assay has a typical lower limit of detection of 0 04 pg/mL for IL-27p28 and 0 03 pg/mL for IL-30
Catalog No. 50-000-78691 Supplier Meso Scale Discovery Supplier No. K152WXD2
The Meso Scale Discovery V-PLEX Mouse IL-27p28/IL-30 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of IL-27p28 and IL-30 in mouse serum plasma and cell culture supernatants The assay uses MSD s proprietary V-PLEX technology is sensitive reproducible and requires a low sample volume of 25 L The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating standard curves The assay has a typical lower limit of detection of 0 04 pg/mL for IL-27p28 and 0 03 pg/mL for IL-30