Learn More
Meso Scale Discovery VPLEX Mouse IL27p28/IL30 Kit
SDP
Catalog No. 5000081864
Encompass
The Meso Scale Discovery V-PLEX Mouse IL-27p28/IL-30 Kit is used to quantitatively measure IL-27p28 and IL-30 in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This multiplex assay kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag and calibrator and control samples It allows concurrent measurement of IL-27p28 and IL-30 from a single sample in as little as 3 5 hours
Catalog No. 50-000-81864 Supplier Meso Scale Discovery Supplier No. K152WXD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Mouse IL-27p28/IL-30 Kit is used to quantitatively measure IL-27p28 and IL-30 in mouse serum plasma and cell culture supernatants It utilizes electrochemiluminescence (ECL) technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This multiplex assay kit includes a plate coated with capture antibodies detection antibodies labeled with an ECL tag and calibrator and control samples It allows concurrent measurement of IL-27p28 and IL-30 from a single sample in as little as 3 5 hours