Learn More
Meso Scale Discovery VPLEX MSE Cytokine 19Plex
SDP
Catalog No. 5000078446
Encompass
The V-PLEX Mouse Cytokine 19-plex Kit from Meso Scale Discovery is a multiplex immunoassay for measuring 19 mouse cytokines and chemokines (e g IL-1 IL-6 TNF- VEGF) from a single sample It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals Suitable for mouse serum plasma tissue culture supernatants and other fluids
Catalog No. 50-000-78446 Supplier Meso Scale Discovery Supplier No. K15255D4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX Mouse Cytokine 19-plex Kit from Meso Scale Discovery is a multiplex immunoassay for measuring 19 mouse cytokines and chemokines (e g IL-1 IL-6 TNF- VEGF) from a single sample It uses electrochemiluminescence detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls buffers and seals Suitable for mouse serum plasma tissue culture supernatants and other fluids