Learn More
Meso Scale Discovery VPLEX Neuroinflammation PNL hu
SDP
Catalog No. 5000082145
Encompass
The Meso Scale Discovery V-PLEX Neuroinflammation Panel 1 Human Kit is used to simultaneously measure biomarkers related to neuroinflammation and neurodegeneration This electrochemiluminescence multiplex assay allows researchers to quantify beta amyloid 1-42 beta amyloid 1-40 total tau and phosphorylated tau in human serum plasma and CSF samples The kit includes a 96-well plate detection antibodies calibrators buffers wash buffers read buffer and plate seals sufficient for one 96-well plate This quantitative immunoassay provides sensitive and accurate results with low sample volume requirements
Catalog No. 50-000-82145 Supplier Meso Scale Discovery Supplier No. K15210D2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Neuroinflammation Panel 1 Human Kit is used to simultaneously measure biomarkers related to neuroinflammation and neurodegeneration This electrochemiluminescence multiplex assay allows researchers to quantify beta amyloid 1-42 beta amyloid 1-40 total tau and phosphorylated tau in human serum plasma and CSF samples The kit includes a 96-well plate detection antibodies calibrators buffers wash buffers read buffer and plate seals sufficient for one 96-well plate This quantitative immunoassay provides sensitive and accurate results with low sample volume requirements