Learn More
Meso Scale Discovery VPLEX NHP Cytokine 24Plex Kit
SDP
Catalog No. 5000081759
Encompass
The V-PLEX NHP Cytokine 24-plex Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures 24 non-human primate cytokines and chemokines in serum plasma and cell culture supernatants The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive plate seals The assay has a typical sensitivity of 0 5-2 pg/mL and a broad dynamic range of up to 4 logs This kit allows for efficient high-throughput quantification of NHP cytokines and chemokines for immunology and inflammation research
Catalog No. 50-000-81759 Supplier Meso Scale Discovery Supplier No. K15058D1
The V-PLEX NHP Cytokine 24-plex Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures 24 non-human primate cytokines and chemokines in serum plasma and cell culture supernatants The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive plate seals The assay has a typical sensitivity of 0 5-2 pg/mL and a broad dynamic range of up to 4 logs This kit allows for efficient high-throughput quantification of NHP cytokines and chemokines for immunology and inflammation research