Learn More
Meso Scale Discovery VPLEX NHP Cytokine 24Plex Kit
SDP
Catalog No. 5000081724
Encompass
The V-PLEX NHP Cytokine 24-plex Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures 24 non-human primate cytokines and chemokines in serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The 24 analytes measured are Eotaxin G-CSF GM-CSF IFN- IL-1 IL-1ra IL-2 IL-4 IL-5 IL-6 IL-8 IL-10 IL-12/23(p40) IL-13 IL-15 IL-17A IL-18 IP-10 MCP-1 MDC MIP-1 MIP-1 TGF- and TNF- This kit allows efficient profiling of non-human primate immune responses
Catalog No. 50-000-81724 Supplier Meso Scale Discovery Supplier No. K15058D2
Add to Cart
Add to Cart
Request a Quote
The V-PLEX NHP Cytokine 24-plex Kit from Meso Scale Discovery is a multiplex immunoassay that simultaneously measures 24 non-human primate cytokines and chemokines in serum plasma and cell culture supernatants The kit uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The 24 analytes measured are Eotaxin G-CSF GM-CSF IFN- IL-1 IL-1ra IL-2 IL-4 IL-5 IL-6 IL-8 IL-10 IL-12/23(p40) IL-13 IL-15 IL-17A IL-18 IP-10 MCP-1 MDC MIP-1 MIP-1 TGF- and TNF- This kit allows efficient profiling of non-human primate immune responses