Learn More
Meso Scale Discovery VPLEX NHP Cytokine 24Plex
SDP
Catalog No. 5000080571
Encompass
The V-PLEX NHP Cytokine 24-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous measurement of 24 non-human primate cytokines and chemokines in serum plasma and culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive plate seals The assay uses electrochemiluminescence for high sensitivity and broad dynamic range The kit can quantify IL-1 IL-1ra IL-2 IL-4 IL-5 IL-6 IL-8 IL-10 IL-12/23(p40) IL-13 IL-15 IL-17A IL-18 Eotaxin G-CSF GM-CSF IFN- IP-10 MCP-1 MIP-1 MIP-1 and TNF-
Catalog No. 50-000-80571 Supplier Meso Scale Discovery Supplier No. K15058D4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX NHP Cytokine 24-plex Kit from Meso Scale Diagnostics is a multiplex immunoassay kit that allows simultaneous measurement of 24 non-human primate cytokines and chemokines in serum plasma and culture supernatants The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and adhesive plate seals The assay uses electrochemiluminescence for high sensitivity and broad dynamic range The kit can quantify IL-1 IL-1ra IL-2 IL-4 IL-5 IL-6 IL-8 IL-10 IL-12/23(p40) IL-13 IL-15 IL-17A IL-18 Eotaxin G-CSF GM-CSF IFN- IP-10 MCP-1 MIP-1 MIP-1 and TNF-