Learn More
Meso Scale Discovery VPLEX NHP GMCSF Kit 25Plate
SDP
Catalog No. 5000081243
Encompass
The V-PLEX NHP GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human GM-CSF It uses electrochemiluminescent assays with patterned arrays and SULFO-TAG labels for high sensitivity and a wide range The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies read buffer wash buffer blocker solutions calibrator and controls Requiring less than 50 L it s suitable for GM-CSF analysis in serum plasma CSF cell culture supernatant and more
Catalog No. 50-000-81243 Supplier Meso Scale Discovery Supplier No. K156RID4
Add to Cart
Add to Cart
Request a Quote
The V-PLEX NHP GM-CSF Kit from Meso Scale Discovery is a multiplex immunoassay for measuring human GM-CSF It uses electrochemiluminescent assays with patterned arrays and SULFO-TAG labels for high sensitivity and a wide range The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies read buffer wash buffer blocker solutions calibrator and controls Requiring less than 50 L it s suitable for GM-CSF analysis in serum plasma CSF cell culture supernatant and more