Learn More
Meso Scale Discovery VPLEX NHP IL12/IL23p4025Plate
SDP
Catalog No. 5000081392
Encompass
The Meso Scale Discovery V-PLEX NHP IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of natural human IL-12 and IL-23 in serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 5-1000 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range
Catalog No. 50-000-81392 Supplier Meso Scale Discovery Supplier No. K156RJD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX NHP IL-12/IL-23p40 Kit is an electrochemiluminescent immunoassay for the quantitative measurement of natural human IL-12 and IL-23 in serum plasma and culture supernatants The kit contains a 96-well plate coated with capture antibodies detection antibodies conjugated to an electrochemiluminescent label calibrators diluent wash buffer read buffer and plate seals The assay has a detection range of 1 5-1000 pg/mL and takes approximately 3 5 hours to complete It utilizes MSD s proprietary SECTOR Imager for high sensitivity and a wide dynamic range