Learn More
Meso Scale Discovery VPLEX NHP IL17A Kit 25Plate
SDP
Catalog No. 5000079935
Encompass
The Meso Scale Discovery V-PLEX NHP IL-17A Kit is an electrochemiluminescent immunoassay for the quantitative measurement of non-human primate interleukin 17A (IL-17A) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of NHP IL-17A with a lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with an IL-17A capture antibody a detection antibody conjugated to an MSD SULFO-TAG calibrators and assay diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation period
Catalog No. 50-000-79935 Supplier Meso Scale Discovery Supplier No. K156RFD4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX NHP IL-17A Kit is an electrochemiluminescent immunoassay for the quantitative measurement of non-human primate interleukin 17A (IL-17A) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible quantitation of NHP IL-17A with a lower limit of detection of 0 04 pg/mL and an upper limit of 10 000 pg/mL The kit includes a 96-well plate coated with an IL-17A capture antibody a detection antibody conjugated to an MSD SULFO-TAG calibrators and assay diluent The assay takes approximately 3 5 hours to complete after a 2 hour incubation period