Learn More
Meso Scale Discovery VPLEX Orthopoxvirus PNL 1 IgG
SDP
Catalog No. 5000081092
Encompass
The Meso Scale Discovery V-PLEX Orthopoxvirus Panel 1 IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to vaccinia virus monkeypox virus cowpox virus and ectromelia virus in serum and plasma samples This multiplex assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide quantitative results with high sensitivity and dynamic range The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals This assay can aid in evaluating orthopoxvirus exposure and immune response
Catalog No. 50-000-81092 Supplier Meso Scale Discovery Supplier No. K15688U4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Orthopoxvirus Panel 1 IgG Kit is an electrochemiluminescence immunoassay that detects IgG antibodies to vaccinia virus monkeypox virus cowpox virus and ectromelia virus in serum and plasma samples This multiplex assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide quantitative results with high sensitivity and dynamic range The kit includes a 96-well plate coated with antigens detection antibodies calibrators controls diluent wash buffer read buffer and plate seals This assay can aid in evaluating orthopoxvirus exposure and immune response