Learn More
Meso Scale Discovery VPLEX Plus Ab Peptide PNL 4G8
SDP
Catalog No. 5000078416
Encompass
The Meso Scale Discovery V-PLEX Plus Abeta Peptide Panel 1 (4G8) Kit is a multiplex immunoassay that measures Abeta38 Abeta40 and Abeta42 in human CSF and plasma samples It uses electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 4G8 antibody that binds to amino acids 17-24 of Abeta It allows quantification of different Abeta peptides which may serve as biomarkers in Alzheimer s disease research and drug development The kit has high lot-to-lot consistency and comes with a plate format of 96 wells
Catalog No. 50-000-78416 Supplier Meso Scale Discovery Supplier No. K15199G2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Abeta Peptide Panel 1 (4G8) Kit is a multiplex immunoassay that measures Abeta38 Abeta40 and Abeta42 in human CSF and plasma samples It uses electrochemiluminescence detection technology and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 4G8 antibody that binds to amino acids 17-24 of Abeta It allows quantification of different Abeta peptides which may serve as biomarkers in Alzheimer s disease research and drug development The kit has high lot-to-lot consistency and comes with a plate format of 96 wells