Learn More
Meso Scale Discovery VPLEX Plus Ab Peptide PNL 4G8
SDP
Catalog No. 5000080628
Encompass
The Meso Scale Discovery V-PLEX Plus Abeta Peptide Panel 1 (4G8) Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Abeta38 Abeta40 and Abeta42 in human CSF and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and specific detection of Abeta peptides The 4G8 antibody enables measurement of total Abeta independent of aggregation state This assay allows researchers to profile Abeta peptides implicated in Alzheimer s disease The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and diluents - everything needed for the immunoassay
Catalog No. 50-000-80628 Supplier Meso Scale Discovery Supplier No. K15199G4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus Abeta Peptide Panel 1 (4G8) Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Abeta38 Abeta40 and Abeta42 in human CSF and plasma It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and specific detection of Abeta peptides The 4G8 antibody enables measurement of total Abeta independent of aggregation state This assay allows researchers to profile Abeta peptides implicated in Alzheimer s disease The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and diluents - everything needed for the immunoassay