Learn More
Meso Scale Discovery VPLEX Plus Ab38 Peptide 4G8
SDP
Catalog No. 5000078312
Encompass
The Meso Scale Discovery V-PLEX Plus A 38 Peptide Kit is designed to quantitatively measure A 38 levels in human samples such as CSF serum and plasma It utilizes electrochemiluminescent detection technology and includes a plate pre-coated with capture antibody specific for the N-terminus of A 38 The kit includes detection antibody calibrator diluent and wash buffer for 96 tests It has high sensitivity and a broad dynamic range to accurately quantify A 38 levels from low pg/mL to high ng/mL concentrations This A 38 assay provides a tool for studying the role of A 38 in Alzheimer s disease pathology
Catalog No. 50-000-78312 Supplier Meso Scale Discovery Supplier No. K150SHG2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery V-PLEX Plus A 38 Peptide Kit is designed to quantitatively measure A 38 levels in human samples such as CSF serum and plasma It utilizes electrochemiluminescent detection technology and includes a plate pre-coated with capture antibody specific for the N-terminus of A 38 The kit includes detection antibody calibrator diluent and wash buffer for 96 tests It has high sensitivity and a broad dynamic range to accurately quantify A 38 levels from low pg/mL to high ng/mL concentrations This A 38 assay provides a tool for studying the role of A 38 in Alzheimer s disease pathology